ESD Safety Clogs, White OB, EASY 2.0 622
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
837622/35 837622/36 837622/37 837622/38 837622/39 837622/40 837622/41 837622/42 837622/43 837622/44 837622/45 837622/46 837622/47 837622/48
ABEB837622/35EA 60.1 EUR
ABEB837622/35 ABEB837622/36 ABEB837622/37 ABEB837622/38 ABEB837622/39 ABEB837622/40 ABEB837622/41 ABEB837622/42 ABEB837622/43 ABEB837622/44 ABEB837622/45 ABEB837622/46 ABEB837622/47 ABEB837622/48
ESD Safety Clogs, White OB, EASY 2.0 622
Schoenen en laarzen
ESD clogs made from grain leather upper material and fabric linning material, with replaceable insole, resistant to acids, bases (cleaning agents).
  • Increased breathability
  • Increased durability of the upper
  • Easy-clean upper
  • Resistance to short-term contact with hot ground at 180 °C
  • Orthopedic insoles used
Suitable for use in ESD environments. Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol.
Colour: White; Fastener type: Buckle.
ESD - protection against electrostatic discharge
Antistatic footwear - antistatic properties
Energy absorption in the heel area - energy absorbtion in the heel area
The sole resistant to diesel fuel - sole resistance to diesel fuel
Certificaten: EN ISO 20347:2012; 120/2023/PPE/1439/B wyd.1; OB (Basic requirements met); SRC (Slip resistance on ceramic).