SGE OptChem™ FocusLiners Inlet Liners
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0926200501 09262005 0926200201
SGEA0926200501EA 57.9 EUR
SGEA0926200501 SGEA09262005 SGEA0926200201
SGE OptChem™ FocusLiners Inlet Liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner. Choosing the right inlet liner and injection parameters can increase peak areas and reduce detection limits by up to 300%.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to use
  • Two focus points in the liner ensures quartz wool remains in the optimum location
  • The wiping of the needle by the quartz wool prevents droplet formation ensuring excellent reproducibility
  • The presence of the quartz wool improves vaporization minimizing high molecular weight discrimination
  • Color coded blue
The SGE FocusLiner® uses a unique design to hold the quartz wool in the correct position.
The FocusLiner is recommended for the following sample or injection modes: General purpose, concentrated samples, dirty samples, split/splitless injection mode.
Bestelinformatie: FocusLiner available for Agilent, Analytical, ATAS, Bruker, GC Inlet FocusLiner, PerkinElmer, Shimadzu GC 2010/17A, Thermo, Thermo Scientific TRACE 1300 Series GC, Varian.