SGE OptChem™ Tapered FocusLiner Inlet Liners
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0926200301 09262003 0926211101 09262111
SGEA0926200301EA 63.5 EUR
SGEA0926200301 SGEA09262003 SGEA0926211101 SGEA09262111
SGE OptChem™ Tapered FocusLiner Inlet Liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner. Choosing the right inlet liner and injection parameters can increase peak areas and reduce detection limits by up to 300%.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to use
  • Bottom taper focuses sample onto the head of the column and minimizes contact with metal parts of the inlet
  • A taper at the top aids in minimizing sample flashback
  • Ensures quartz wool remains in the correct position in the liner
  • Excellent reproducibility results from the wiping of the sample from the syringe needle and the prevention of droplet formation
  • Color coded aqua
The Tapered FocusLiner® is recommended for the following sample or injection modes: Trace level analysis, splitless, dirty sample, wide boiling point range.
Bestelinformatie: Tapered FocusLiner® available for Agilent, Analytical, Bruker, GC Inlet FocusLiner, GC Inlet Tapered FocusLiner, PerkinElmer, Shimadzu, Shimadzu GC 2010/17A, Thermo Scientific TRACE 1300 Series GC, Varian.