SGE OptChem™ Straight Inlet Liners
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0926200101 09262001 0926200701 09262007 0926201601 09262016
SGEA0926200101EA 38.3 EUR
SGEA0926200101 SGEA09262001 SGEA0926200701 SGEA09262007 SGEA0926201601 SGEA09262016
SGE OptChem™ Straight Inlet Liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner. Choosing the right inlet liner and injection parameters can increase peak areas and reduce detection limits by up to 300%.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to use
  • Straight liners facilitate higher split flows
  • Narrow bore straight liners facilitate fast GC work
  • Small injection volumes of less than 0,5 µl are best used
  • Narrow bore straight liners improve focusing of gaseous samples (purge, trap and headspace)
  • Color coded purple
Straight liners are recommended for the following sample or injection modes: General purpose, concentrated samples, dirty samples (only if quartz wool is present), gaseous samples, purge and trap, headspace, split/splitless.
Bestelinformatie: For Agilent, Analytical, ATAS, Bruker, GC Inlet FocusLiner, PerkinElmer, Shimadzu GC 2010/17A, Thermo, Thermo Scientific TRACE 1300 Series GC, Varian