Reach-In Refrigerator with Bottom Compressor, FRFCvg
Leverancier: Liebherr
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FRFCVG5501 FRFCVG6511 FRFCVG5511
LIEBFRFCVG5501EA 0 EUR
LIEBFRFCVG5501 LIEBFRFCVG6511 LIEBFRFCVG5511
Reach-In Refrigerator with Bottom Compressor, FRFCvg
Koelkasten
This FRFCvg Reach-In refrigerator with bottom compressor save time and money. In case of an uncontrolled temperature rise, if a door has been left open or there were any other problems, the interior light of the appliance will start flashing.
  • Environmentally friendly refrigerants
  • Efficient refrigeration system
  • Interchangeable door hinge
  • Display lock
  • Mechanical lock
  • Easy to clean interior
  • Grid shelf with pull-out stop
  • Digital temperature display
  • Reversible door hinges (left/right)
This refrigerators for the catering industry are generally designed for use with Euro standard gastronomy containers. From an opening angle of approx. 90°, the appliance door remains open and facilitates stocking and unloading as well as cleaning. Below approx. 90°, it closes by itself so that the appliance does not remain open accidentally.
It is with the retrofittable WiFi and LAN interface, appliances can be effortlessly connected to external systems. If the power supply was interrupted due to a fault, the power failure information appears once the power is restored.