DURAN® Glass Tubes
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DR19/1.9AR DR38/2.0AR 1028437 DR40/2.4AR 322.0AR 1485594 1544262 1544261 1552469 1580606 1580607 1580608 1580609 1621123 1637429 1637432 1397497 1529018 1662270 1552466 1028561 1667897 1643569 1716790 1716791 1716799 1716852 1027516 1027813
SCORDR19/1.9AREA 0 EUR
SCORDR19/1.9AR SCORDR38/2.0AR SCOR1028437 SCORDR40/2.4AR SCOR322.0AR SCOR1485594 SCOR1544262 SCOR1544261 SCOR1552469 SCOR1580606 SCOR1580607 SCOR1580608 SCOR1580609 SCOR1621123 SCOR1637429 SCOR1637432 SCOR1397497 SCOR1529018 SCOR1662270 SCOR1552466 SCOR1028561 SCOR1667897 SCOR1643569 SCOR1716790 SCOR1716791 SCOR1716799 SCOR1716852 SCOR1027516 SCOR1027813
DURAN® Glass Tubes
Slangen Transfer Tubing
High precision glass tubes made from AR® glass.
  • Good chemical and physical properties
  • Not resistant to high thermal loads or rapid temperature changes
DURAN® Glass tubes are suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals.