SamplePro Needle Mounting Adapter
Leverancier: Bruker
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRUKH160098EA 183 EUR
BRUKH160098
SamplePro Needle Mounting Adapter
Injectienaalden
SPT NADELINSERT KPL is required to mount new needle version in the SamplePro Tube liquid Handler.
Suitable for needles H160071, H160072, H160073. One insert per SamplePro Tube required.