Heat Resistant Gloves
Leverancier: JUTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H115AS238-W2 H115AS230-W2
JUTEH115AS238-W2EA 187 EUR
JUTEH115AS238-W2 JUTEH115AS230-W2
Heat Resistant Gloves
Handschoenen
Five finger glove made of glass fabric with silicone coating and preox-aramide fabric, aluminized.
  • Abrasion-resistant, non-slip
  • Also available with additional lining made of preox-fleece
  • Also available with hook-and-loop closure
  • Flexible, excellent handling
  • Material: Glass fabric up to 600 °C with silicone coating (palm)
Certificaten: EN 388:2016 (4X4XX), EN 407 (433444).