Aluminized Heat Protection Coats
Leverancier: JUTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HSM120KA-1-54 HSM120KA-1-56
JUTEHSM120KA-1-54EA 647 EUR
JUTEHSM120KA-1-54 JUTEHSM120KA-1-56
Aluminized Heat Protection Coats
Werkkleding
Coat made of preox-aramide fabric heat protection coat.
  • Flexible fabric
  • Very pleasant wearing comfort
  • High heat reflection
  • Insulated snaps, stand-up collar, Velcro fastener
  • Weight: 260 g/m²
Certificaten: EN ISO 11612:2015.
Order Now
Specificaties
Environmentally Preferable
Kledingstuk toepassing Industrial
Risk area Heat
Geslacht
Sleeve or leg length Long