VWR® Crimp Neck Vials ND11
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
548-2645AEA 1020 EUR
548-2645A 548-2646A
VWR® Crimp Neck Vials ND11
flesjes
Standard vials for GC and HPLC.
  • 1st hydrol. class, label and filling lines
  • Top bonded