Torque Wrench
Leverancier: CEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CEMA514140EA 0 EUR
CEMA514140
Torque Wrench
Moersleutels
Torque wrench, for use with all EasyPrep Vessels.
All wrenches are calibrated, preset and wrapped with acid resistant chemical coating for optimum vessel performance, repeatability and longevity.
Using a non-calibrated wrench may lead to poor results, reduce vessel longevity and corrosion of the wrench.
Bestelinformatie: Wrench does not come with the required socket, can be ordered separately.