Filter holder with funnel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS0310-4000 DS0310-4050
513-3101EA 213 EUR
513-3101 NALGDS0310-4050
Filter holder with funnel, Nalgene®
Filterhouders
These graduated polysulfone filter holders can be used with any filtering flask or manifold that accepts a suitable rubber stopper with hole.
  • Environmentally friendly alternative to disposable filters
Filtration area, with analytical support plate: 11,25 cm².
Filtration area, with sterilisation support plate: 13,30 cm².
Leveringsinformatie: Comes with one easy-to-use, no twist vacuum gasket which can replace a No. 7 or 8 rubber stopper with 15 mm hole.