Spare burettes, Dr. Schilling Pattern
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
23713 23715 23716 23718 23723 23724
612-3269EA 49.7 EUR
612-3269 BRND23715 612-3270 612-3271 BRND23723 BRND23724
Spare burettes, Dr. Schilling Pattern
Buretten
AR® glass, class B.
  • Calibrated to deliver 'Ex'
  • Schellbach stripe, reduced distance between subdivision marks
DIN EN ISO 385
Bottle requirements: 500 ml for 10 and 15 ml; 1000 ml for 25 and 50 ml
Certificaten: DIN EN ISO 385.