Analogue stopwatches, Amigo
Leverancier: Hanhart
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Amigo
609-1240EA 178 EUR
609-1240
Analogue stopwatches, Amigo
Timers Analoge stopwatches
Analogue stopwatches with robust ABS casing.
  • Pin lever, one jewel
  • ¹/₁₀ second
  • 15 minutes capacity
  • Shock-protected pin lever movement
  • Robust ABS case, with cord for hanging round the neck