iFluor® 405, Succinimidyl Ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1021 71021 71501 71551
AATB1021EA 307 EUR
AATB1021 AATB71021 AATB71501 AATB71551
iFluor® 405, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Opgelet: For research use only.