D(-)-Luciferin sodium salt monohydrate ≥95%
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
12509 12510 12511
AATB12509EA 120 EUR
AATB12509 AATB12510 AATB12511
D(-)-Luciferin sodium salt monohydrate ≥95%
D-(-)-Luciferin sodium salt monohydrate
Luciferin is the most popular and versatile bioluminescent substrate.
Formule: C₁₁H₇N₂NaO₃S₂·H₂O
MW: 320.32 g/mol
MDL Nummer: MFCD11865368
CAS nummer: 103404-75-7
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9545cea070c2a9fa67002828fa7e826c6e81e881&vid=4acb507e8831cd7b26a5d8d8d9b4ebd3ac5d9a55&prid=web&clid=SAPDAM