FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
13417 13418
AATB13417EA 307 EUR
AATB13417 AATB13418
FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
Activation of caspases plays a central role in apoptosis.
  • Caspase detection
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5 II