Fluo-8® potassium salt
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
21087 21089
AATB21087EA 460 EUR
AATB21087 AATB21089
Fluo-8® potassium salt
Fluo-8® potassium salt
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms
Opgelet: For research use only.
MW: 877.07 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=474814bcaa14bf5672850ad5254b1834eee60e6c&vid=767cca80f96500707ddb7a229247ce469580f97d&prid=web&clid=SAPDAM