Cross linked-AlloPhycocyanin fluorescent probe
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
CL-APC
2549
2550
2551
2552
AATB2549EA
537
EUR
AATB2549
AATB2550
AATB2551
AATB2552
Cross linked-AlloPhycocyanin fluorescent probe
Cross linked-AlloPhycocyanin
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
- High extinction coefficient
- High quantum efficiency
- Large Stokes shift
MW:
105000 g/mol |
ADR:
5 II |