RNAintact Stabilisation Solution, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11RINTA050 11RINTA100 11RINTA005
11RINTA050EA 100.8 EUR
11RINTA050 11RINTA100 11RINTA005
RNAintact Stabilisation Solution, MP Biomedicals
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
RNAintact stabilisation solution is an aqueous sample storage reagent that stabilizes and preserves RNA in samples for extended period at a wide range of storage conditions (−80 to +37 °C).
  • Quick and easy-to-use RNA preservation buffer
  • Format: Liquid
  • Product type: RNA preservation solution
  • Reagent storage ambient temperature: 15 to 25 °C
  • Applications: Refrigerated (4 °C) >1 month
  • Sample stability: Bacteria, tissue culture cells, animal tissues, plant tissues
The solution has been tested for compatibility with various sample types, ranging from bacteria, tissue culture cells, animal tissues and even preservation of some plant tissues.
By storing samples in RNAintact stabilisation solution, the solution protects the RNA from degradation by rapidly inactivating the enzymatic activities of RNases for an RNase-free storage environment.
It is easy to perform RNA isolation from samples stored in RNAintact stabilisation solution, as the solution is compatible with most RNA isolation procedures, including TRIzol™ and spin column kits. Samples stored in RNAintact stabilisation solution yield high quality RNA, suitable for downstream applications (e.g. gene expression by PCR and realtime PCR).