Amplite® Fluorimetric Malondialdehyde (MDA) Quantitation Kit
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10072 10071
AATB10072EA 614 EUR
AATB10072 AATB10071
Amplite® Fluorimetric Malondialdehyde (MDA) Quantitation Kit
Testen Enzyme-assays
Malondialdehyde (MDA) is one of the natural byproducts during lipid peroxidation.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Opgelet: For research use only.