MMP Green™ Substrate
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13519 13520
AATB13519EA 307 EUR
AATB13519 AATB13520
MMP Green™ Substrate
Testen Enzyme-assays
The matrix metalloproteinases (MMPs) constitute a family of zinc-dependent endopeptidases that function within the extracellular matrix.
  • Bright signal
  • Consistent results
  • In-house quality testing
Opgelet: For research use only.