HEX-dT Phosphoramidite
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6210 6211
AATB6210EA 281 EUR
AATB6210 AATB6211
HEX-dT Phosphoramidite
Koppelings- en labelingssystemen voor eiwitten
HEX-dT phosphoramidite provides a valuable tool to add the HEX tag to a desired location of an oligo sequence.
  • DMT group allows quantification
HEX-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue.
It is complimentary to the commonly used 6-HEX phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.
Verpakking: Plastic vial.
Opgelet: For research use only.