5-VIC Phosphoramidite
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6083 6084 6085
AATB6083EA 997 EUR
AATB6083 AATB6084 AATB6085
5-VIC Phosphoramidite
Koppelings- en labelingssystemen voor eiwitten
VIC phosphoramidite is the most convenient building block to incorporate VIC tag into an oligonucleotide.
  • Emits in the green-yellow part of the visible spectrum
  • Multiple applications
VIC is a fluorescent dye that was originally developed by Applied Biosystems (now, Thermofisher). It is used to fluorescently label oligonucleotides at the 5’-end (for use as probes in a variety of real-time PCR, hybridization and fluorescence-based genetic analysis applications).
Verpakking: Plastic vial.
Opgelet: For research use only.