ReadiLink™ Cy5 Oligo and ssDNA Labeling Kit
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17488 17489
AATB17488EA 353 EUR
AATB17488 AATB17489
ReadiLink™ Cy5 Oligo and ssDNA Labeling Kit
Koppelings- en labelingssystemen voor eiwitten
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
  • Robust protocol
  • Bright labeling
The resulting Cy5-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Verpakking: Packaged in plastic vials.
Opgelet: For research use only.