Covipyte™ ED450
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13537 13538
AATB13537EA 292 EUR
AATB13537 AATB13538
Covipyte™ ED450
Eiwitten en peptiden
Covidyte™ ED450 is a peptide substrate containing 12 amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Dabcyl (quencher) and Edans (donor) on the C-and N-terminals respectively where the fluorescence of Edans is effectively quenched by Dabcyl when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the Edans fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by Dabcyl. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of Edans. Covidyte™ ED450 is a convenient tool for screening inhibitors of coronavirus proteases.
  • MW 2034.36
Opgelet: For research use only.