MagaDye™ 613 ddCTP
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17060 17064
AATB17060EA 1410 EUR
AATB17060 AATB17064
MagaDye™ 613 ddCTP
Reagentia voor nucleïnezuren Reagentia voor sequentiebepaling
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Opgelet: For research use only.