Mini molecular models: DNA
Leverancier: SPIRING ENTERPRISES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MDNA-060-11 MDNA-060-22
763-0453EA 0 EUR
763-0453 763-0454
Mini molecular models: DNA
Modellen Moleculaire biologie modellen
Mini molecular models of DNA with nucleic acids, five-sided sugar molecules and pyramid-shaped phosphate molecules.
Only correct base pairs can be connected.
Bestelinformatie: Components include colour-coded bases: G, C, A, T.