Ice Cubers, Self Contained, IM
Leverancier: HOSHIZAKI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IM-100CNE-HC23 IM-100CNE-HC
HOSHIM-100CNE-HC23EA 4130 EUR
HOSHIM-100CNE-HC23 HOSHIM-100CNE-HC
Ice Cubers, Self Contained, IM
IJsmakers
Self contained cube ice makers producing high-quality cube ice. Meeting even the strictest hygiene requirements, this true allrounder offers many application possibilities in segments including the foodservice and medical sector.
  • Uses refrigerant R290, this natural refrigerant is not only climate-friendly, but also boosts the machine, when it comes to performance and production capacity
  • The exterior is made of stainless steel
  • The closed water circuit provides maximum protection against any type of contamination during the entire cubing process
  • Magnetic water pump without direct coupling, prevents leakage from the water circuit and improves the life expectancy of the machine
  • Perfect fit for small spaces
  • Easy to use, clean, and maintain
  • The smart plug-and-play design guarantees an easy and quick set-up
All Cubers are equipped with an electronic control system. This ensures an optimised ice-making process, even under varying circumstances. Without additional manual assistance or adjustments required, produces generous amounts of ice cubes, at a consistently high quality.
A dedicated jet of fresh water is injected into each of the closed cells. Excess water leaves the closed cell through a set of outlets. The cell walls block the freezing water’s expansion from all sides while reaching down to –30 °C during the cubing process. Thanks to the automatic rinse cycles, impurities such as minerals, are eliminated from the water resulting in the purest ice possible while your machine is protected from calcification and mineral build-up.
Power supply: 1/220 - 240 V/50 Hz; Electric consumption: 0,5 kW; Heat rejection: 1,15 kW (for large and medium cubes), 2,33 kW (for extra small cubes).
Cooling system: Air cooled; CO₂ equivalent: 0,375 kg.
Gross weight packed: 88 kg.