Azijnzuur-D1 (98% D) for NMR spectroscopy
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Ethanoic acid-D1
166200500 166202500
ACRO166200500EA 87.8 EUR
ACRO166200500 ACRO166202500
Azijnzuur-D1 (98% D) for NMR spectroscopy
Azijnzuur-D1
Formule: C₂H₃DO₂
MW: 61.04 g/mol
Kookpunt: 116…117 °C (760 mmHg)
Smeltpunt: 15…16 °C
Dichtheid: 1.07 g/cm³
Vlampunt: 40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051050
CAS nummer: 758-12-3
EINECS: 212-059-1
UN: 2789
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤1 % (Coulometric)
D-Enrichment ≥98.5 %