Instrument Baskets with Perforated Sides
Leverancier: HUPFER METALLWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7500809 7500645 7515007 7500599
HUPF7500809EA 219 EUR
HUPF7500809 HUPF7500645 HUPF7515007 HUPF7500599
Instrument Baskets with Perforated Sides
Manden Draadmanden
Immersion baskets for transport, storage and provision of various sterile supply instruments.
  • Produced from electro-polished stainless steel
  • Mesh size 6×6 mm
  • Perforated plate thickness 0,8 mm
  • Without stackable frame
Baskets in hygienic design for easy cleaning, made of high-quality, electro-polished stainless steel round material, with walls made of finely perforated plates and base made of welded mesh. Easy handling thanks to two interior chest handles at the long sides.