ProLite™ His-Tag Protein Gel Staining Kit
Leverancier: AAT Bioquest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ProLite™
AATB18010EA 546 EUR
AATB18010
ProLite™ His-Tag Protein Gel Staining Kit
Eiwitexpressie Reagentia voor eiwitexpressie
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
  • Green fluorescence
  • Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.
Opgelet: For research use only.