ReadiLink™ Cy5 Nick Translation dsDNA Labelling Kit
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17409 17408
AATB17409EA 682 EUR
AATB17409 AATB17408
ReadiLink™ Cy5 Nick Translation dsDNA Labelling Kit
Koppelings- en labelingssystemen voor eiwitten
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted Cy5-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridisation (FISH)..
Verpakking: Packaged in plastic vials.
Opgelet: For research use only.