Buccutite™ Rapid PE-Cy5.5 Tandem Antibody Labeling Kits (2 Labellings)
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1316
1341
AATB1316EA
769
EUR
AATB1316
AATB1341
Buccutite™ Rapid PE-Cy5.5 Tandem Antibody Labeling Kits (2 Labellings)
Koppelings- en labelingssystemen voor eiwitten
PE-Cy5.5 is a popular colour used in flow cytometry.
- High conjugation yield
- Stable crosslinking
- Minimal hands-on time
- Robust protocol