Screen Quest™ Membrane Potential Assay Kits, Red Fluorescence
Leverancier: AAT BIOQUEST
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36004
36005
36006
AATB36004EA
307
EUR
AATB36004
AATB36005
AATB36006
Screen Quest™ Membrane Potential Assay Kits, Red Fluorescence
Testen
Cellulaire assays
Membrane potential is the difference in voltage between the interior and exterior of a cell.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Opgelet: For research use only.