PhosphoWorks™ Luminometric ATP Assay Kit, DTT-free
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21612 21613
AATB21612EA 230 EUR
AATB21612 AATB21613
PhosphoWorks™ Luminometric ATP Assay Kit, DTT-free
Testen Cellulaire assays
Adenosine triphosphate (ATP) plays a fundamental role in cellular energenics, metabolic regulation and cellular signaling.
  • Extreme sensitivity
  • Stable signal with ~4 hour half-life
  • No mixing or separations required
  • No DTT used
Opgelet: For research use only.