Stayright™ Purple, ReadiUse™ HRP Chromogen Premixed with Hydrogen Peroxide*
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45900 45901
AATB45900EA 307 EUR
AATB45900 AATB45901
Stayright™ Purple, ReadiUse™ HRP Chromogen Premixed with Hydrogen Peroxide*
Stayright™ Purple
3,3'-Diaminobenzidine (DAB) has been used as the most commonly used IHC chromogen because it is inexpensive and sensitive for routine applications.
  • Ready to use
  • Non-mutagenic
  • Minimal cytotoxicity
  • No mixing steps required
  • Stain persists through dehydration and coverslipping
Opgelet: For research use only.