Streptavidin-Xtra™ IF488
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
46000 46001
AATB46000EA 152 EUR
AATB46000 AATB46001
Streptavidin-Xtra™ IF488
Eiwitten en peptiden
Streptavidin conjugates are widely used in combing with biotin conjugates for detecting a variety of biological targets such as proteins, nucleic acids and other molecules.
  • Improved signal
  • Improved photostability
  • Multiple applications
Opgelet: For research use only.