Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 405)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16524 16771
AATB16524EA 152 EUR
AATB16524 AATB16771
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 405)
Antilichamen
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Opgelet: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse