iFluor® 350, Succinimidyl Ester fluorescent probe
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1020 71020 71500 71550
AATB1020EA 307 EUR
AATB1020 AATB71020 AATB71500 AATB71550
iFluor® 350, Succinimidyl Ester fluorescent probe
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Opgelet: For research use only.