iFluor® 670, Succinimidyl Ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71033 71512 71562 1033
AATB71033EA 114 EUR
AATB71033 AATB71512 AATB71562 AATB1033
iFluor® 670, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Opgelet: For research use only.