iFluor® 568, Succinimidyl Ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1049 71049 71522 71572
AATB1049EA 307 EUR
AATB1049 AATB71049 AATB71522 AATB71572
iFluor® 568, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Opgelet: For research use only.