6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥95%
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 6-FAM SE, 6-Carboxyfluorescein NHS ester, 6-Carboxyfluorescein N-hydroxysuccinimide ester, 6-FAM-N-succinimidyl ester
116 117 118
AATB116EA 152 EUR
AATB116 AATB117 AATB118
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥95%
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)
6-FAM, SE is the amine-reactive succinimidyl ester of single isomer 6-FAM acid.
Formule: C₂₅H₁₅NO₉
MW: 473.39 g/mol
MDL Nummer: MFCD00077323
CAS nummer: 92557-81-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=075c29e63190b5c01b15c0d616d98ecf6fe5cdf9&vid=dc1eacf64c004320f7c83449b710c3e253a1873e&prid=web&clid=SAPDAM