FITC-C6-LEHD-FMK
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
13407 13409
AATB13407EA 769 EUR
AATB13407 AATB13409
FITC-C6-LEHD-FMK
FITC-C6-LEHD-FMK
Activation of caspases plays a central role in apoptosis.
  • Caspase detection
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0e8831c7ef32e7d06809d27028777fb15a11891&vid=5a1005b725b201725b725423939830800c187146&prid=web&clid=SAPDAM