5-FAM-YVAD-FMK ≥90%
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13473 13483
AATB13473EA 152 EUR
AATB13473 AATB13483
5-FAM-YVAD-FMK ≥90%
5-FAM-YVAD-FMK
FAM-YVAD-FMK, i.e., 5-carboxyfluorescein-Tyr-Val-Ala-Asp-fluoromethylketone, binds irreversibly to active caspase 1 in stimulated cells.
  • Bright Fluorescent Signal
  • Specific Caspase 1 Binding
Opgelet: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7809b6a81d50e79ceef7a86a882f7da5df51ca9&vid=02fb05db231c58df2a77a8635acba873c6fa4df7&prid=web&clid=SAPDAM