iFluor® 665, Reactive Form_Succinimidyl Ester fluorescent probe
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
1550 1551 1552 1553
AATB1550EA 137 EUR
AATB1550 AATB1551 AATB1552 AATB1553
iFluor® 665, Reactive Form_Succinimidyl Ester fluorescent probe
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • pH-insensitive
  • High photostability
  • Brighter signal
ADR: 5 II