Dextran, M.W. 3,000 Cal-590™ labelled
Leverancier: AAT BIOQUEST INC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AATB20508EA 442 EUR
AATB20508
Dextran, M.W. 3,000 Cal-590™ labelled
Dextran
Calcium measurement is critical for numerous biological investigations.
  • Well-retained in neurons
  • 3,000 mw
  • Useful for in vivo imaging
From Eukaryotic cells
Opgelet: For research use only.
Formule: (C₆H₁₀O₅)n
MDL Nummer: MFCD00130935
CAS nummer: 9004-54-0