Cal Red™ R525/650 AM
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
20590 20591
AATB20590EA 769 EUR
AATB20590 AATB20591
Cal Red™ R525/650 AM
Cal Red™ R525/650 AM
The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels.
  • Calcium-dependent fluorescence
  • Improved sensitivity
  • Compatible with hts filters
  • Ratiometric detection
  • Cell permeant
Opgelet: For research use only.
ADR: 5,II