Calbryte™ 590 potassium salt
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
20706 20707
AATB20706EA 460 EUR
AATB20706 AATB20707
Calbryte™ 590 potassium salt
Calbryte™ 590 potassium salt
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein & 'coupled receptors' (GPCRs) and calcium channel targets.
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell impermeant
Opgelet: For research use only.
MW: 1074.98 g/mol
ADR: 5 II