Sulpho-Cyanine 3 NHS ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
263 264
AATB263EA 769 EUR
AATB263 AATB264
Sulpho-Cyanine 3 NHS ester
Sulpho-Cyanine 3 NHS ester
A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • pH Tolerance From 3 - 10
  • Bright Fluorescence
  • Multiple Bioconjugation Applications
Opgelet: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b53d38e0a6f60c17a1e38747285bbd0c85d58f88&vid=65e08f6eb9bf46035c8e5b604c5727eef3d101d2&prid=web&clid=SAPDAM